About this item
Pico™ (ventilated) and Fresco™ (refrigerated) centrifuges are available in two speed options. Pico™ and Fresco™ 17 are perfect for routine laboratory work, while the Pico™ and Fresco™ 21 are ideal for high-end research applications.
- The 24-place rotor can be used for all 1,5 to 2,0 ml tubes, including mini-preps and spin columns
- Composite rotor technology, for greater chemical resistance and faster acceleration/deceleration compared to traditional aluminium
- Unique ClickSeal® biocontainment rotor lid 'clicks' in place, providing assurance that samples are securely contained, and with one click opening, shortens retrieval time compared to screw-on lids
- PPSU transparent lids have improved strength and chemical resistance compared with polycarbonate/metallic lids, and enables viewing of the samples for added safety and convenience
- Intuitive controls and easy to read display simplifies operation; IEC 61010-1, IEC 61010-2-020
Ideal for everyday laboratory use, Thermo Scientific Pico 17 microcentrifuges provide outstanding processing power, unique safety features and a broad array of microcentrifuge rotors and accessories for microcentrifuge tube processing.
Accelerate routine sample preparation processes up to 17,000×g with our standard microcentrifuge, available with cooling. When our Thermo Scientific Fresco 17 is coupled with our extensive microcentrifuge rotor selection, these microcentrifuges offer productivity, versatility, safety and convenience in a compact, easy-to-use lab instrument.
Support micro-volume protocols such as nucleic acid or protein lysate preparation and PCR reaction set-up all in a remarkably small footprint with Thermo Scientific Pico 21 microcentrifuges. Choose between seven lightweight microcentrifuge rotors, designed to provide acceleration up to 21,100×g in as little as 12 seconds.
Specifications
- Catalog No:
- HERA75002420
- HERA75002425
- HERA75002475
- HERA75002476
- HERA75002491
- HERA75002410
- 型号:
- Fresco™ 17
- Fresco™ 21
- Pico™ 21
- Fresco™ 21
- Pico™ 17
- Pico™ 17
- 最大容量:
- 24×1,5/2,0 ml
- 24×1,5/2,0 ml
- 24×1,5/2,0 ml
- 24×1,5/2,0 ml
- 24×1,5/2,0 ml
- 24×1,5/2,0 ml
- 最大速度:
- 13300 min⁻¹
- 14800 min⁻¹
- 14800 min⁻¹
- 14800 min⁻¹
- 13300 min⁻¹
- 13300 min⁻¹
- 最大相对离心力:
- 17000 ×g
- 21100 ×g
- 21100 ×g
- 21100 ×g
- 17000 ×g
- 17000 ×g
- 温度:
- –9...+40 °C (in increments of 1 °C)
- –9...+40 °C (in increments of 1 °C)
- –9...+40 °C (in increments of 1 °C)
- 加速/制动时间:
- 12/12 s
- 12/12 s
- 12/12 s
- 12/12 s
- 12/12 s
- 12/12 s
- 计时器:
- 1 - 99 min (in increments of 1 min) + Hold
- 1 - 99 min (in increments of 1 min) + Hold
- 1 - 99 min (in increments of 1 min) + Hold
- 1 - 99 min (in increments of 1 min) + Hold
- 1 - 99 min (in increments of 1 min) + Hold
- 1 - 99 min (in increments of 1 min) + Hold
- 噪音电平:
- 50 dB(A)
- 50 dB(A)
- 56 dB(A)
- 50 dB(A)
- 56 dB(A)
- 56 dB(A)
- 电气:
- 230 V 50/60 Hz
- 230 V 50/60 Hz
- 230 V 50/60 Hz
- 230 V 50/60 Hz
- 230 V 50/60 Hz
- 230 V 50/60 Hz
- 宽度×深度×高度:
- 295×445×330 mm
- 295×445×330 mm
- 243×352×225 mm
- 295×445×330 mm
- 243×352×225 mm
- 243×352×225 mm
- 重量:
- 28 kg
- 28 kg
- 11 kg
- 28 kg
- 11 kg
- 11 kg