pH electrode solutions
02, Pepsin cleaning solution, 250 ml, PEP/pH, 3
供应商: WTW
Jump to:
Product Details & Documents
Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted.
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.
规格
规格
- 包装规格:3
- 说明:Pepsin cleaning solution, 250 ml, PEP/pH
- 顺序:02