pH micro electrodes, InLab® Micro / InLab® Nano / InLab® NMR
目录编号 METT51343161
供应商: METTLER TOLEDO
规格
- 连接:S7
- 直径×长度:6×230 mm
- 用于:Highly saline solutions; cosmetics; pharmaceutical and biological samples; dairy products; water-based paint and oily samples
- Type:InLab® Semi-Micro-L
- 说明:InLab® Semi-Micro-L
- 材质:玻璃
- 顺序:04
- 隔膜:Ceramic
- 电解质:3 mol/l KCl
- 包装规格:1
- 温度:0...+100 °C
- pH:0 - 14
规格
关于此项目
pH electrodes with S7 or BNC connector.
- ARGENTHAL™ reference system with silver-ion trap (except InLab® Nano / NMR: Ag/AgCl)
- XEROLYT® EXTRA polymer electrolyte: High signal stability, very short response time and high chemical resistance (InLab® Semi-Micro)
- InLab® Flex-Micro features a flexible thin shaft (Ø 6 mm): Ideal for measuring pH in test tubes and other small containers
pH range: 0 - 14 for InLab® Micro/Micro Pro/Semi-Micro-L; 0 - 12 for InLab® Semi-Micro
Material: Glass
Ordering information: Supplied with test certificate. Other electrodes available on request.