Orders LinkContactUsLinkComponent
Frio temp精密温度计
Frio temp精密温度计
   可持续选项
供应商:   VWR Collection
Frio temp精密温度计
供应商:  VWR Collection
   可持续选项
Certificates
Frio temp精密温度计
10 Options Available Below
Product Details & Documents
该款玻璃温度计量取冷却器、冰箱、培养箱、烤炉和血库的温度。
  • Enviro safe non toxic and liquid or red spirit liquid
  • Accurate temperature reading without sample contamination
  • Individual serial number
  • Wide choice
Accurate temperature reading is achieved by using the filled bottle (included), which mimics the sample. Insert the thermometer into the bottle and place near the samples for best results. PFA coating safely contains glass and liquid in case of breakage.
Traceable to NIST/DKD
Certifications: NIST可追溯性。
Delivery information: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability plus VELCRO® and magnetic attachments.
Specifications for Product Family
规格
  • Catalog No:
  • VWRI620-0854
  • VWRI620-0902
  • VWRI620-0848
  • VWRI620-0878
  • VWRI620-0855
  • VWRI620-0843
  • VWRI620-0845
  • VWRI620-0856
  • VWRI620-0844
  • VWRI620-0886
  • Environmentally Preferable:
  • Safer Human and Environmental Health
规格
Product Family Options
Enviro safe
产品信息
  • 顺序
    说明
    范围
    准确度
    分度
    长度
    填充
    可用库存
    Price
  • 01
    冷却器用
    −30…+1 °C
    ±0.5 °C
    0,5 °C
    135 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,459.00
    ¥1,459.00
    1 * 1 件
       ¥1,459.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0854
    Environmentally Preferable
  • 03
    冰箱用
    −5...+15 °C
    ±0.5 °C
    0,5 °C
    125 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,613.00
    ¥1,613.00
    1 * 1 件
       ¥1,613.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0902
    Environmentally Preferable
  • 04
    血库用
    −5...+20 °C
    ±0.5 °C
    0,5 °C
    135 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,443.00
    ¥1,443.00
    1 * 1 件
       ¥1,443.00
    规格:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    VWRI620-0848
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • 05
    培养箱用
    +15…+50 °C
    ±0.5 °C
    0,5 °C
    135 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,585.00
    ¥1,585.00
    1 * 1 件
       ¥1,585.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0878
    Environmentally Preferable
红水
产品信息
  • 顺序
    说明
    范围
    准确度
    分度
    长度
    填充
    可用库存
    Price
  • 09
    用于超低温冰箱
    −90...+20 °C
    ±1 °C; −1/−40 °C为±2 °C; −40 °C以下为±2,5 °C
    1,0 °C
    145 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,807.00
    ¥1,807.00
    1 * 1 件
       ¥1,807.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0855
    Environmentally Preferable
  • 10
    冷却器用
    −30...+1 °C
    ±0.5 °C
    0,5 °C
    135 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,852.00
    ¥1,852.00
    1 * 1 件
       ¥1,852.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0843
    Environmentally Preferable
  • 11
    冰箱用
    −5...+15 °C
    ±0.5 °C
    0,5 °C
    125 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,724.00
    ¥1,724.00
    1 * 1 件
       ¥1,724.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0845
    Environmentally Preferable
  • 13
    培养箱用
    +15…+50 °C
    ±0.5 °C
    0,5 °C
    135 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,386.00
    ¥1,386.00
    1 * 1 件
       ¥1,386.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0856
    Environmentally Preferable
  • 14
    烤炉用
    +20…+130 °C
    ±1.0 °C
    1,0 °C
    145 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥1,320.00
    ¥1,320.00
    1 * 1 件
       ¥1,320.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0844
    Environmentally Preferable
  • 15
    烤炉用
    +50…+220 °C
    ±2.0 °C
    2,0; 3,0 (>130) °C
    145 mm
    Orange/yellow liquid
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥2,384.00
    ¥2,384.00
    1 * 1 件
       ¥2,384.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI620-0886
    Environmentally Preferable