Nalgene™ Staining Boxes
01, Staining boxes, PMP, 500 ml, 127×127×508 mm
NALG5705-1010
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods.
- Polymethylpentene box and polypropylene copolymer cover
- Useful over a –70 to +70 °C (–94 to +158 °F) temperature range
- Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
- LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
- Plug won't interfere with cover or contents of boxes on a benchtop or shaker
- Stackable, with a handle on each end for safe, easy carrying
- Microwavable and easy to clean
Thermo Scientific™ Nalgene™ staining box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 189×189 mm (4,8×4,8") gel along with 500 ml of liquid.
- 容量:500 ml
- 说明:Staining boxes
- 材质:PMP
- 顺序:01
- 宽度×深度×高度:127×127×508 mm
- Cat. No.:NALG5705-1010
- 环境认证:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact