Beta radiation shields, Nalgene®
Catalog # NALG6700-1812
Supplier: Thermo Fisher Scientific
Specifications
- 形状:Angled
- 宽度×深度×高度:305×305×457 mm
- Environmentally Preferable:Low Manufacturing Impact
- 环境认证:NAR Certificate of Retirement
- 包装规格:1
Specifications
About this item
Acrylic, transparent, thickness: 9 mm.
- The deep base (305 mm) provides a large flat work surface and minimises the risk of 'hot spots' on the lab bench
- Designed to protect the lab worker when handling beta emitting isotopes
Meets OSHA Standard 29 CFR Part 1910, 1030
Certifications: Meets OSHA Standard 29 CFR Part 1910, 1030