Robotic tips, Axygen®, for FLIPR Liberty 384 (Type B)
供应商: Corning
证书
关于此项目
Transparent PP.
- Sterile tips: Sterilised by gamma radiation
- Filter tips: PE filter prevents any pipette contamination
- Free of RNase, DNase, DNA and pyrogens
规格
- Catalog No:
- AXYGLT-384-R
- Environmentally Preferable:
- Low Manufacturing Impact
- 环境认证:
- I-RECs, ISO 14001 - Reynosa