ContactUsLinkComponent
Pipette controller, accu-jet® pro
Pipette controller, accu-jet® pro
Catalog # BRND26310
Supplier:  BRAND
undefined
Pipette controller, accu-jet® pro
Catalog # BRND26310
Supplier:  BRAND
Specifications
  • 颜色代码:
    深蓝色
  • 说明:
    accu-jet® pro, with battery charger for UK (230 V/50 Hz)
  • 顺序:
    09
  • 包装规格:
    1
Specifications
About this item
Pipette controller for glass and plastic pipettes from 0,1 to 200 ml.
  • One handed operation of delivery mode, gravity delivery or blow-out, and continuous adjustment of filling/delivery speed using variable button pressure
  • Intelligent electronic system prevents overloading
  • Flashing LED indicates 2 hours in advance when the battery needs charging
  • Silently aspirates 50 ml in one go, in just 10 seconds
  • Operates for up to 8 hours with 10 ml pipette
  • Can be kept in the wall support or placed on its back to keep the pipette away from work surfaces
Weight: 190 g
Ordering information: Supplied with a rechargeable NiMH battery, two battery compartment covers, wall support, battery charger, two spare 0,2 µm membrane filters.