Pasteur pipettes, VOLAC®
01, 6,8 - 7,1 mm, 150 mm, -, Bulk
Catalog # POULD810
Supplier: POULTEN GRAF
Jump to:
Product Details & Documents
Glass, graduated, plain or pre-plugged, sterile or non sterile.
- Precision pipettes with a heavy wall thickness to provide the best quality product allied with greater strength to give lower breakages during transit, and whilst in use or, when fitting PVC teats
- Standard pipettes with top body, outer diameter 6,8 to 7,1 mm, wall thickness 0,50 to 0,53 mm and jet outer diameter 1,50 ±0,2 mm
Documents
Specifications
- 外径:6,8 - 7,1 mm
- 包装规格:Bulk
- 灭菌:-
- 长度:150 mm
- 顺序:01
Accessories
Product Information
- Catalog #说明AvailabilityPrice