Transfer pipettes
08, Graduated, 3,0 ml, 40 µl, 150 mm, -, Bulk
目录编号 RATI2600111
供应商: RATIOLAB
Jump to:
Product Details & Documents
PE, disposable.
- Soft, elastic bulb
- Unbreakable, reduced risk of injury compared to glass
- Inert to biological fluids and most acids
- Graduated pipettes with clearly visible embossed graduations
- Sterile pipettes are gas-sterilised and individually packed
- Suitable for food contact
规格
- 包装规格:Bulk
- 灭菌:-
- 容量:3,0 ml
- 说明:Graduated
- 长度:150 mm
- 顺序:08
- 液滴体积:40 µl