Orders LinkContactUsLinkComponent
Mops, PolyGen™
Mops, PolyGen™
   Sustainable Options Available
Supplier:   MICRONOVA
Mops, PolyGen™
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Mops, PolyGen™
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The dual-action abrasive mesh is specially adapted to pharmaceutical and biotech projects. Heads used directly with the fibreglass/PP handle or attached to the universal handle via the quick disconnect adapter.
  • Mop can clean to a dry finish or apply disinfectants requiring extended drying time
  • Special herringbone weave ensures no raw edges to guarantee low particulation
  • Low-linting polyester looped strands
  • Reusable, launderable, and autoclavable
Ordering information: Mop heads and handle must be ordered separately.
Specifications for Product Family
Specifications
  • Catalog No:
  • MICRPG9-12
  • MICRPG9-12IR
  • Environmentally Preferable:
  • Waste Reducing
Specifications
Product Family Options
Product Information
  • 说明
    Availability
    Price
  • Mop head
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRPG9-12
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • Mop head, sterile
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MICRPG9-12IR
    Environmentally Preferable
Accessories
Product Information
  • Catalog #
    顺序
    说明
    Availability
    Price
  • MICRQDMH-1
    04
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    Log in for availability
    1 * 1 件
Product Information
  • Catalog #
    顺序
    说明
    长度
    Availability
    Price
  • MICRSSU-1
    01
    不锈钢可伸缩手柄
    965 - 1829 mm