Mops, MicroMop™
Product Details & Documents
MicroMop™ are ideal for quick and efficient cleaning of large surface areas. Suitable for wafer processing, electronics, pharmaceutical, and biotech applications. All mop heads attach to stainless steel universal handle via the orange polypropylene quick disconnect adapter. For less critical applications, choose the fibreglass/polypropylene handle with attached adapter.
- Low-linting, quick drying PVA is ideal for high-tech applications. Can be laundered
- Polyester has good wicking capability, can be autoclaved and used with strong disinfectants
- Lite™ is sturdy latex/rayon blend for controlled environments and cleanroom support areas; mop head can be autoclaved for medical and sterile applications
: All components must be ordered separately.
规格
- 说明:Mop head, NovaLite™
- 顺序:02
- Cat. No.:MICRCRM-2
- Environmentally Preferable:Waste Reducing
Accessories
产品信息
- 目录编号顺序说明可用库存Price
- MICRQDMH-104MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesLog in 查看库存1 * 1 件