Chamber slides, Nunc™ Lab-Tek™ II CC2
Supplier: Thermo Fisher Scientific
Certificates
About this item
Borosilicate coverglass. PS chamber. Sterile.
- Growth surface remains stable without refrigeration
- Light blue frosted writing area for clear identification
- Slide separator to lift the medium chamber from the slide in each pack
Chemically coated growth surface on the glass slide, which mimics poly-lysine, providing binding sites for fastidious cells such as neurons.
Specifications
- Catalog No:
- NUNC154941
- NUNC154917
- NUNC154739
- NUNC154852
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- 环境认证:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement