PCR plates, 96-well, segmented, Axygen®
AXYGPCR-96-SG-C
: Corning
- 孔数量:96
- 裙边:半裙边
- Well colour:透明
- Environmentally Preferable:Low Manufacturing Impact
- 环境认证:I-RECs, ISO 14001 - Reynosa
- 包装规格:10
- 孔体积:320 μl
PP plates, silicone mats. Designed to fit 0,2 ml thermal cycler blocks.
- Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
- Certified free from DNase, RNase and endotoxin for highest sample integrity
- AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 240 μl
Compatible with (in addition to standard thermal cycling): BioRad iCycler, MyiQ, iQ5, Beckman CEQ sequencers.