Plates, Nunc™ OmniTrays
关于此项目
PS, sterile, with lid. Useful for screening libraries and for DNA hybridisation methods, these 90 ml trays are also suitable for standard Petri dish techniques and bacterial growth.
- Compatible with robotic systems
- Sturdy wall design facilitates handling and stacking
- Can be frozen at −20 °C
External dimensions: 128×86 mm
Recommended working volume: 35 ml
Recommended working volume: 35 ml
2 Options Available Below
Product Details & Documents
PS, sterile, with lid. Useful for screening libraries and for DNA hybridisation methods, these 90 ml trays are also suitable for standard Petri dish techniques and bacterial growth.
- Compatible with robotic systems
- Sturdy wall design facilitates handling and stacking
- Can be frozen at −20 °C
External dimensions: 128×86 mm
Recommended working volume: 35 ml
Recommended working volume: 35 ml
Specifications for Product Family
规格
- Catalog No:
- NUNC242811
- NUNC264728
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- 环境认证:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
规格
Product Family Options
产品信息
- 说明灭菌可用库存Price
- 规格:
Low Manufacturing Impact More
Specifications
Cat. No.NUNC242811Environmentally PreferableLow Manufacturing Impact环境认证NAR Certificate of RetirementLow Manufacturing Impact - 规格:
Low Manufacturing Impact More
Specifications
Cat. No.NUNC264728Environmentally PreferableLow Manufacturing Impact环境认证NAR Certificate of RetirementLow Manufacturing Impact