Orders LinkContactUsLinkComponent
Staining systems, EasyDip™
Staining systems, EasyDip™
供应商:   SIMPORT SCIENTIFIC
Staining systems, EasyDip™
供应商:  SIMPORT SCIENTIFIC
Certificates
Staining systems, EasyDip™
Staining systems, EasyDip™
9 Options Available Below
Product Details & Documents
The slide staining system is made of acetal plastic.
  • Jars can be loosely joined to each other to keep the same order when moved around
  • Wide base for stability while inside is recessed to reduce reagent volume
  • Available in five colours to aid identification of contents or applications
The slide staining system has two components: A square staining jar and a 12 position vertical slide rack. The staining jar is made of resistant acetal plastic and will resist most staining agents including alcohol and xylene but not phenol. Autoclavable, it is therefore suitable for special stains, frozen sections and special processes.
Volume: 80 ml
W×D×H: 76×64×92 mm
Ordering information: The staining jars and staining rack are sold separately, except for the assorted package (720-0791), which contains one jar of each colour (blue, green, pink, white, yellow) and a rack. The EasyDip aluminum holder is designed to hold six EasyDip slide staining jars. The EasyDip slide staining kit (SIMPM906-12AS) contains 6 jars (blue, green, pink, 2xwhite, yellow), an anodized aluminium rack, and one slide staining rack.
Product Family Options
产品信息
  • 顺序
    说明
    颜色
    材质
    宽度×深度×高度
    可用库存
    Price
  • 01
    Slide staining rack and five jars
    组合
    乙缩醛
    76×64×92 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 KIT
    1 * 1 KIT
  • 02
    Slide staining cuvettes
    蓝色
    乙缩醛
    76×64×92 mm
    Log in 查看库存
    Log in 查看库存
    1 * 6 件
    1 * 6 件
  • 03
    Slide staining cuvettes
    绿色
    乙缩醛
    76×64×92 mm
    Log in 查看库存
    Log in 查看库存
    1 * 6 件
    1 * 6 件
  • 04
    Slide staining cuvettes
    粉红色
    乙缩醛
    76×64×92 mm
    Log in 查看库存
    Log in 查看库存
    1 * 6 件
    1 * 6 件
  • 05
    Slide staining cuvettes
    白色
    乙缩醛
    76×64×92 mm
    Log in 查看库存
    Log in 查看库存
    1 * 6 件
    1 * 6 件
  • 06
    Slide staining cuvettes
    黄色
    乙缩醛
    76×64×92 mm
    Log in 查看库存
    Log in 查看库存
    1 * 6 件
    1 * 6 件
  • 07
    Vertical slide staining racks
    深灰色
    乙缩醛
    76×64×92 mm
    Log in 查看库存
    Log in 查看库存
    1 * 6 件
    1 * 6 件
  • 08
    EasyDip™ slide staining kit
    组合
    乙缩醛
    76×64×92 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
Holder for EasyDip
产品信息
  • 顺序
    说明
    颜色
    材质
    宽度×深度×高度
    可用库存
    Price
  • 09
    Holder (for up to six jars)
    不锈钢
    425×102×38 mm