Orders LinkContactUsLinkComponent
Tissue grinders, conical
Tissue grinders, conical
01, 1 ml, 11×49 mm, 130 mm
目录编号 WHEA358103
供应商:   DWK LIFE SCIENCES
Tissue grinders, conical
目录编号 WHEA358103
供应商:  DWK LIFE SCIENCES
Tissue grinders, conical
Tissue grinders, conical
01, 1 ml, 11×49 mm, 130 mm
Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents
Borosilicate glass. Handheld homogeniser with conical surface, which allows initial size reduction, followed by passage through the cylindrical section for the final homogenisation step. Grinding efficiency is improved and less time is required as compared to the Tenbroeck and Potter-Elvehjem designs. Recommended for heart and lung tissues, as well for skin and plant tissues.
  • Clearance between pestle and tube is 0,1 to 0,15 mm
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifications: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
规格
  • 容量:
    1 ml
  • 顺序:
    01
  • 总长度:
    130 mm
  • 外径×长度:
    11×49 mm