Cleanroom gloves, nitrile, Kimtech™ G3 NxT™
供应商: KIMBERLY CLARK
证书
关于此项目
Ambidextrous gloves with beaded cuffs. Contain no natural rubber latex.
- Non sterile, powder-free
- Textured fingertips to facilitate control and grip
- Static dissipative in use
- Increased cleanliness
Suitable for Class 10, ISO 4, M2.5 cleanrooms.
Length: 300 mm
Thickness: 0,13 mm (palm)
Colour: White
Certifications: EN 374-1 Type C (K), EN 374-5 (VIRUS), Cat. III; AQL 1.5
Packaging: Packaged in cleanroom compatible polyethylene, 100 gloves per double poly bag, 10 bags per pack.
规格
- Catalog No:
- KIMB62991
- KIMB62990
- KIMB62992
- 左右手通用的/分左右手:
- 左右手通用的
- 左右手通用的
- 左右手通用的
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- 一次性/可重复使用:
- 一次性
- 一次性
- 一次性
- 有/无粉:
- 无粉
- 无粉
- 无粉
- 洁净室等级:
- 1/ISO 3
- 1/ISO 3
- 1/ISO 3
- 袖口形式:
- 珠状
- 珠状
- 珠状
- 袖口厚度:
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 手指厚度:
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 掌心厚度:
- 0,13 mm
- 0,13 mm
- 0,13 mm
- 欧洲标准:
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- Anti-static:
- +
- +
- +