Orders LinkContactUsLinkComponent
Funnels with extra-long stem
Funnels with extra-long stem
Supplier:   KARTELL
Funnels with extra-long stem
Supplier:  KARTELL
Certificates
Funnels with extra-long stem
Funnels with extra-long stem
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PP, transparent.
  • Inner ribs to accelerate the filtration process
  • Very good chemical resistance, autoclavable
  • Suitable for use with standard filter papers
With inner ribs for rapid filtration. Cone angle 60°.
Documents
Product Family Options
Product Information
  • 杆长度
    杆外径
    漏斗外径
    Availability
    Price
  • 150 mm
    8 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 143 mm
    8 mm
    78,5 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 145 mm
    8 mm
    103,5 mm