Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
镊子
Forceps with guide pin
Forceps with guide pin
Forceps with guide pin
Supplier:
BOCHEM
Certificates
Expand image
Forceps with guide pin
Supplier:
BOCHEM
About this item
Stainless steel, with PTFE coating.
Bent, with guide pin
Smooth, coated surface to prevent contamination and facilitate cleaning