Volumetric flasks, class A
06, 1000 ml, 24/29, 0,40 ± ml
目录编号 VITL67604
供应商: VITLAB
Jump to:
Product Details & Documents
PMP (TPX®), transparent, with PP stopper.
- Individually calibrated ring mark
- Imprinted lot number
- Complete with certificate
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
规格
- 模拟器:24/29
- 容量:1000 ml
- 顺序:06
- 误差:0,40 ± ml