Volumetric flasks, class B
1000 ml, 19/26, 0,80 ± ml
KART2489
: KARTELL
PMP (TPX®), transparent, with stopper.
- Autoclavable
- For working temperatures up to +170 °C
- With NS stoppers
- 模拟器:19/26
- 容量:1000 ml
- 误差:0,80 ± ml
PMP (TPX®), transparent, with stopper.