ContactUsLinkComponent
Flasks, round bottom with three necks and standard ground joints
Flasks, round bottom with three necks and standard ground joints
Catalog # LENZ03314158
Supplier:  LENZ LABORGLAS
undefined
Flasks, round bottom with three necks and standard ground joints
Catalog # LENZ03314158
Supplier:  LENZ LABORGLAS
Specifications
  • 外径:
    105 mm
  • 容量:
    500 ml
  • 顺序:
    16
  • 包装规格:
    1
  • 侧管颈:
    14/23 NS
  • 中心颈:
    29/32 NS
Specifications
About this item
DURAN®, borosilicate glass, with angled side necks, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck and two side necks, both inclined at an angle of 20°. Matt white labelling area.
Cork rings for round bottom flasks
Product Information