Erlenmeyer flasks, graduated with SVL screw caps
BIBB1137/08D
- 外径:105 mm
- 螺纹:SVL 42
- 容量:500 ml
- 高度:170 mm
- 顺序:07
- 包装规格:1
PYREX®, borosilicate glass, clear, with black phenolic screw caps with PTFE faced liner.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
White graduations and white labelling area.