Syringe filters, Acrodisc® ion chromatography (IC)
Optimised to provide the most consistent results when analysing ionic species. Certified for low level extractables detected by conductivity.
Supor® (hydrophilic polyethersulfone); membrane PP housing.
Acrodisc® PSF syringe filters are Zymark and SOTAX Automation Certified to ensure smooth operation and worry-free performance 24 hours a day in automated workstations.
- Catalog No:
- PALL4585
- PALLAP-4785
- PALL4785
- PALL4483
- PALL4485
- PALL4683
- PALL4685
- PALLAP-4585
- PALL4783
- PALL4583
- 类型:
- Syringe filter
- Premium syringe filter (PSF)
- Syringe filter
- Syringe filter
- Syringe filter
- Syringe filter
- Syringe filter
- Premium syringe filter (PSF)
- Syringe filter
- Syringe filter
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- 直径:
- 25 mm
- 25 mm
- 25 mm
- 13 mm
- 13 mm
- 13 mm
- 13 mm
- 25 mm
- 25 mm
- 25 mm
- 孔大小:
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,20 µm
- 0,45 µm
- 0,20 µm
- 0,45 µm
- 0,45 µm
- 0,20 µm
- 0,20 µm
- 最大运行压力:
- -
- 4,1 bar at 21 − 24 °C; 2,1 bar at 100 °C
- -
- 6,9 bar
- 6,9 bar
- 6,9 bar
- 6,9 bar
- 4,1 bar at 21 − 24 °C; 2,1 bar at 100 °C
- -
- -
- 最高温度:
- -
- 100 °C
- -
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 100 °C
- -
- -
- 水流速:
- -
- 420 ml/min (2,1 bar)
- -
- 15 ml/min (1 bar)
- 20 ml/min (1 bar)
- 15 ml/min (1 bar)
- 20 ml/min (1 bar)
- 420 ml/min (2,1 bar)
- -
- -