Dissection set
Dissection set, 8 pcs.
目录编号 BOCH12017
供应商: BOCHEM
Jump to:
Product Details & Documents
Stainless steel. This dissection set is designed for academic applications. Includes.
- One pair of scissors with two sharp cutting tips
- One pair of scissors with a sharp and a blunt cutting tip
- Two scalpels
- Two forceps
- Two dissection needles
规格
- 说明:Dissection set, 8 pcs.