Incinerating dishes
Product Details & Documents
Porcelain dishes, glazed inside and out, except base.
- Withstand maximum temperature 1000 °C
- Shallow form without spout, deep form with spout
The rectangular dishes have sloping sides.
Measurements shown are W×D×H for rectangular dishes and Ø×H for others.
规格
- Type:Flour (Mohs)
- 容量:30 ml
- 顺序:07
- 宽度×深度×高度:- mm
- 外径×高度:56×23 mm