Orders LinkContactUsLinkComponent
Storage pots
Storage pots
1100 ml, 199,7 mm, 190 mm, 147 mm, 89 mm
Catalog # KART143
Supplier:   KARTELL
Storage pots
Catalog # KART143
Supplier:  KARTELL
Storage pots
Storage pots
1100 ml, 199,7 mm, 190 mm, 147 mm, 89 mm
Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PP, white.
  • Very good chemical and temperature resistance
  • Robust
  • Impermeable to liquids
Ideal for keeping instruments, pipettes, thermometers and many other laboratory tools tidy and close to hand. With extra wide base for stability.
Specifications
  • 外径:
    147 mm
  • 内径:
    89 mm
  • 容量:
    1100 ml
  • 内高:
    190 mm
  • 外部高度:
    199,7 mm