Orders LinkContactUsLinkComponent
Faeces containers
01, 15 ml, 非无菌, 22 mm, 63 mm
  KART600
 :   KARTELL
Faeces containers
  KART600
 :  KARTELL
Faeces containers
Faeces containers
01, 15 ml, 非无菌, 22 mm, 63 mm
1 * 1.000 件
PS, transparent. Non sterile or sterile.
  • With PP spatula incorporated into the cap
  • 直径:
    22 mm
  • 无菌:
    非无菌
  • 容量:
    15 ml
  • 高度:
    63 mm
  • 顺序:
    01