Anion exchange HPLC columns, RCX-10
RCX-10 carbohydrate analysis columns are designed for the isocratic or gradient separation of carbohydrates. When an isocratic method is used with a conductivity, refractive index, ultraviolet, or pulsed amperometric detector (PAD), mono- and di-saccharides such as glucose, fructose, sucrose and lactose can be quickly determined.
- Greater exchange capacity than that of the PRP-X100
- Samples with two or three carbohydrates can be quickly separated isocratically
- More complex samples require gradient elution
- Catalog No:
- HAMI79379
- HAMI79463
- HAMI79440
- HAMI79462
- HAMI79378
- HAMI79388
- 最大压力:
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- pH 范围:
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 温度限值:
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)