Specifications
- 列格式:Capillary
- 薄膜厚度:0,40 µm
- 内径:0,32 mm
- 说明:on 7" cage
- 长度:50 m
- 顺序:25
- Phase:CP-Wax 52 CB
- 包装规格:1
Specifications
About this item
The CP-Wax™ lines of chemically bonded columns offer a broad range of selectivities for method optimisation. The backbone of this series stationary phases is made of a polyethylene glycol.