SGE BPX5 GC Capillary Columns
目录编号 SGEA054125
Supplier: Trajan Scientific and Medical
Specifications
- 列格式:Capillary
- 薄膜厚度:0,5 µm
- 内径:0,32 mm
- 长度:25 m
- 顺序:21
- Polarity:Low-polarity stationary phase
- 温度限值:370 °C
- Phase:BPX5
- 包装规格:1
Specifications
About this item
The SGE BPX5 GC Capillary Column provides a great low-polarity option for GC analyses. Recommended for: Ultra trace analyses, pesticides, herbicides, hydrocarbons, solvents, phenols, amines.
- High temperature
- General-purpose GC column (suitable for over 80% routine GC analyses)
- Very low bleed – ideal for trace analysis
- Non-polar
- Extremely inert
- Ideal for GC-MS
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.