Cleanroom face masks, Kimtech™ M3, PE
Catalog # KIMB62467
Supplier: KIMBERLY CLARK
Specifications
- Type:With ties
- 颜色:白色
- 顺序:2
- 尺寸:18 cm
- 包装规格:200
Specifications
About this item
Sterile gamma-irradiated face masks suitable for use in Class 1, ISO 3, M1.5 environments.
- Outer layer of breathable PE film protects from contamination
- Sterile polyester binding and polyester filter media
- Bacteria filtration efficiency of ≥89% and particle filtration efficiency of ≥90%
Packaging: Individually packed. Packs are double bagged.