Automatic burettes, Dr. Schilling pattern, Schellbach
04, 25 ml, 0,1 ml, 330 mm, 0,045 ± ml
Catalog # HIRS3600172
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Borosilicate 3.3 glass, silicone tubing, PVC fitting, PE bottle and PP base.
- Schellbach stripe, blue graduations
- Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
With bottle: 500 ml for 5 and 10 ml; 1000 ml for 15, 25 and 50 ml.
Documents
Specifications
- 容量:25 ml
- 顺序:04
- 分度:0,1 ml
- 误差:0,045 ± ml
- 刻度长度:330 mm