Carboys, wide neck, PP, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PPCO, translucent with white PP screw cap.
- Very good chemical resistance
- Heat resistant up to 121 °C, autoclavable
- Extra wide neck, wide handles on the shoulders, leakproof, graduated
Ideal for use in microbiology, perfect for handling large quantities of liquids, powders or other solids and pastes.
Specifications
- Catalog No:
- NALG2235-0020
- NALG2235-0050
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- 环境认证:
- NAR Certificate of Retirement
- NAR Certificate of Retirement