Beakers, Philips
01, 250 ml, 105 mm, 58 mm
目录编号 BIBB1020/06D
供应商: DWK LIFE SCIENCES
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Conical shape with spout and labelling area.
规格
- 外径:58 mm
- 容量:250 ml
- 高度:105 mm
- 顺序:01