Size exclusion chromatography columns, OHpak SB series
SHOWF6429106
: Resonac
- 列格式:Analytical
- 流速:1,2 ml/min
- 内径:8,00 mm
- 颗粒大小:13 µm
- 孔大小:15000 Å
- 最大压力:3,0 MPa (30 bar)
- 说明:OHpak SB-806M HQ
- 长度:300 mm
- 美国药典代码:L38, L39
- pH 范围:3 - 10
- 温度限值:4 - 70 °C
- 排除极限:20000000* (Pullulan)
- 硬件栏:Stainless steel
- 包装规格:1
SB-800 HQ are polyhydroxymethacrylate-based SEC columns for molecular weight distribution analysis of water-soluble polymers. The solvent can be substituted with DMF (except for SB-802 HQ and SB-807 HQ columns), enabling the analysis of polar polymers.
- Polymer-based SEC (GFC) columns for aqueous systems
- Support a wide range of molecular weight
- SB-807 HQ with an exclusion limit of 500000000 is available for the analysis of macromolecules
* Estimated value