亮蓝R
Glass bottle for solids, 10 g
目录编号 BRAD161-0400
Marketsource
供应商: BIO RAD
Jump to:
Product Details & Documents
Coomassie Brilliant Blue R-250 protein stain powder
同义词: Coomassie 亮蓝R
- CAS号:6104-59-2
- 化学式:C₄₅H₄₄N₃NaO₇S₂
- MDL 编号:MFCD00041762
- Molecular Weight:825.98 g/mol
- 熔点:174…180 °C
- 储存温度:Ambient
- EINECS:228-060-5
规格
规格
- 包装类型:Glass bottle for solids
- 包装规格:10 g



