亮蓝R solution
5037, Glass bottle, 1 L
Catalog # BRAD161-0436
Marketsource
Supplier: BIO RAD
Jump to:
Product Details & Documents
Use this Coomassie brilliant blue R-250 solution to stain proteins in SDS-PAGE gels.
- Ready to use for fast and easy staining
- Mixture of water, methanol, and glacial acetic acid
同义词: Coomassie 亮蓝R
- CAS号:6104-59-2
- 化学式:C₄₅H₄₄N₃NaO₇S₂
- MDL 编号:MFCD00041762
- Molecular Weight:825.98 g/mol
- 熔点:174…180 °C
- 储存温度:Ambient
- EINECS:228-060-5
- UN:2924
- ADR:3 (8),III
规格
规格
- 包装类型:Glass bottle
- 包装规格:1 L
- 顺序:5037



