Specifications
- 说明:EasySep™ mouse CD90.2 positive selection kit II, 2×10⁹ cells
- 顺序:01
- 包装规格:1 KIT
Specifications
About this item
Immunomagnetic positive selection of CD90.2+ cells from mouse splenocytes or other tissues.
- Fast and easy-to-use
- Up to 98% purity
- No columns required
- Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD90.2 Positive Selection Kit II is designed to isolate CD90.2+ (Thy1.2+) cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognising CD90.2 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Mouse CD90.2 Positive Selection Kit for even faster cell isolations and does not result in the labelling of isolated cells with PE.