Specifications
- 说明:EasySep™ mouse hematopoietic progenitor cell isolation kit, 1×10⁹ cells
- 顺序:01
- 包装规格:1 KIT
Specifications
About this item
Immunomagnetic isolation of untouched mouse stem/progenitor cells
- Fast and easy-to-use
- Up to 84% purity
- No columns required
- Untouched, viable cells
The EasySep™ Mouse Hematopoietic Cell Isolation Kit is designed to isolate stem and progenitor cells from single-cell suspensions of bone marrow or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-hematopoietic stem cells and non-progenitor cells (CD5, CD11b, CD19, CD45R/B220, Ly6G/C(Gr-1), TER119, 7-4) and streptavidin-coated magnetic particles (RapidSpheres™ ). labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
This product replaces the EasySep™ Mouse Hematopoietic Progenitor Cell Enrichment Kit (Catalog #19756) for faster cell isolations.