Specifications
- 说明:EasySep™ mouse MDSC (CD11b+Gr1+) isolation kit, 1×10⁹ cells
- 包装规格:1
Specifications
About this item
Immunomagnetic isolation of untouched myeloid-derived suppressor cells (CD11b+Gr1+).
- Fast, easy-to-use and column-free
- Up to 96% purity
- Untouched, viable cells
The EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit is designed to isolate CD11b+Gr1+ myeloid-derived suppressor cells (MDSCs) from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-MDSCs and streptavidin-coated magnetic particles (RapidSpheres™). labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, cell sorting, and cell culture.