VWR® Stackable mini rollers, designed for gentle agitation of multiple sized bottles/tubes in a space saving footprint.
- Adjustable digitial speed control, timer
- Sealed electronics, constructed of corrosion resistant materials, suitable for use in CO₂/95% humidity environments
- Remote control (wired) included, to control unit without disrupting CO₂ Incubator chamber environment
- 3D swing model offers 18 mm swing motion
- Easy to add and remove rollers to accommodate a wide variety of tube/bottle sizes
- Optional stacking kit to stack 2 units, maximising bench space
All models include O-rings for 50 ml tubes and 15 ml tubes for smooth rolling and wired remote control module.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- VWRU76670-460
- VWRU76670-462
- 最大容量:
- 6,5 kg, 3×1500 ml bottles
- 6,5 kg, 3×1500 ml bottles
- 速度范围:
- 0,5 - 80 rpm
- 0,5 - 80 rpm
- 尺寸:
- 360×378×72 mm; Roller (Ø×L): 28×269 mm
- 360×378×72 mm; Roller (Ø×L): 28×269 mm