Thermo Scientific™ Nalgene™ Two- and Three-Way Stopcocks with Hose Barb Connections
Supplier: MASTERFLEX PREMIUM
Certificates
About this item
Easily adjust flow.
- Tubulated outlets are arranged in a T shape and accepts 1/4 through 7/16" tubing
- Polypropylene, with non-seizing, Teflon™ TFE plug
- Autoclavable
Experience in-house fluid transfer system flexibility with the two- and three-way stopcock, arranged in a straight (two-way) or T-shape (three-way) to connect any two — or all three — arms of the stopcock with ease
- Environmentally Preferable:
Specifications
- 连接类型:
- Barbed
- Barbed
- Barbed
- Barbed
- Inlet port:
- 1/2" to 5/16" Barb
- 1/2" to 5/16" Barb
- 1/2" to 5/16" Barb
- 1/4" to 5/16" Barb
- Outlet port:
- 1/2" to 5/16" Barb
- 1/2" to 5/16" Barb
- 1/2" to 5/16" Barb
- 1/4" to 5/16"
- 内管直径:
- 0,25 - 0,3125"
- 0,25 - 0,3125"
- 0,25 - 0,3125"
- 0,25 - 0,3125"
- 孔口:
- 2 mm (0,0787")
- 4 mm (0,157")
- 2 mm (0,0787")
- 4 mm (0,157")
- 质保:
- 1 year
- 1 year
- 1 year
- 1 year
- 浸湿材料:
- Polypropylene
- Polypropylene
- Polypropylene
- Polypropylene