- 包装规格:5000/pack
- 灭菌:+
- 版本:Graduated
- 容量:1,5 ml
- 颜色:自然色
- 材质:PP
- 最大相对离心力:26,000×g
Improve sample recovery by significantly reducing surface binding with Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes. Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. With no additives or coatings.
- Manufactured from virgin polypropylene, these microcentrifuge tubes ensure sample integrity during boiling, freezing and centrifugation
- Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
- Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
The duckbill cap reduces the pressure required to open the tube with a wider contact surface, yet ensures the cap will remain closed during centrifugation. Low retention properties are not achieved by a coating or plastic-embedding processes, ensuring that samples will not be contaminated from these additives. Tubes will remain closed during centrifugation yet open easily with one hand as needed. Tubes are available in amber to protect your light-sensitive samples. Graduation marks on the microcentrifuge tube allow for effective sample reference.
A variety of packaging options are available to help with contamination control in the lab. For stringent sterility requirements, individually wrapped sterile tubes are available. Color options, in addition to natural and amber, are red, blue, green, yellow and violet and a mixed color packaging.