About this item
ICP-MS multi element solution standard at 10 µg/ml with 12 month expiration date.
Solution A of the 3 solution set ICP-MS-68A with 48 components:
Aluminum, arsenic, barium, beryllium, bismuth, boron, cadmium, calcium, cerium, cesium, chromium, cobalt, copper, dysprosium, erbium, europium, gadolinium, gallium, holmium, indium, iron, lanthanum, lead, lithium, lutetium, magnesium, manganese, neodymium, nickel, phosphorus, potassium, praseodymium, rhenium, rubidium, samarium, scandium, selenium, sodium, strontium, terbium, thallium, thorium, thulium, uranium, vanadium, ytterbium, yttrium, zinc at 10 µg/ml in 2% HNO₃
Solution B of the 3 solution set ICP-MS-68A with 13 components:
Antimony, germanium, hafnium, molybdenum, niobium, silicon, silver, tantalum, tellurium, tin, titanium, tungsten, zirconium at 10 µg/ml in 2% HNO₃ + Tr HF
Solution C of the 3 solution set ICP-MS-68A with 7 components:
Gold, iridium, osmium, palladium, platinum, rhodium, ruthenium at 10 µg/ml in 15% HCl