ContactUsLinkComponent
Combined ion-selective electrodes, perfectION™
Combined ion-selective electrodes, perfectION™
Catalog # METT51344718
Supplier:  METTLER TOLEDO
undefined
Combined ion-selective electrodes, perfectION™
Catalog # METT51344718
Supplier:  METTLER TOLEDO
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • 浓度:
    5×10-8 - 1 mol/l
  • 连接:
    BNC, 1,2 m cable
  • 说明:
    perfectION™ I-
  • 材质:
    环氧树脂
  • 顺序:
    13
  • 电解质:
    D
  • 包装规格:
    1
  • 温度:
    0...+80 °C
  • pH:
    0 - 12
Specifications
About this item
Combined ion-selective electrodes consisting of a measuring electrode complete with an integrated reference.
  • Click & Clear™ junction enables optimum contact between the electrolyte and measurement solution, even in difficult samples
  • The electrolyte solution can be made to flow easily through the junction by pressing the electrode head downwards
  • The junction needs to be cleaned between measurements and the electrolyte can be simply emptied and refilled if necessary
Other electrodes available on request. For further information please contact your local Avantor office.
ISA solution for Na+: NH₄Cl/NH₄OH
Ordering information: Each electrode is supplied with one replacement module. The reference electrolytes must be ordered separately.