Specifications
- 说明:Set for micropipette FX 5/FX 10
- 包装规格:1
Specifications
About this item
Spare/replacement part set for the micropipette FX 5/FX 10 series pipettors.
Set includes 1 cone, 2 PTFE seals, 2 O-rings, and 1 cone nut.