Glass Inserts for IM3 Microscopes
目录编号 OPTKIM3GLASS
供应商: OPTIKA
规格
- 说明:Glass insert for IM3 microscope
- 包装规格:1
规格
关于此项目
Inverted microscopes are useful for observing living cells or organisms at the bottom of a large container (e.g., a tissue culture flask) under more natural conditions than on a glass slide, as it occurs with a conventional microscope. IM-3 series is engineered and designed to be your ideal solution for fast and reliable routine inspections, with the exclusive, state-of-the-art X-LED8 illumination system.
A particularly simple and ingenious optical design allows stable alignments and smooth and accurate movements.
OPTIKA provides different configurations, including the innovative LED fluorescence technology for a new, enhanced
experience.