VWR® Inoculating Loops and Needles, Sterile
Catalog # VWRU76544-932
Supplier: VWR BRAND PRIVATE LABEL USA
Specifications
- 包装规格:25/pack
- 容量:—
- 颜色:白色
- 说明:Needles
- 顺序:08
Specifications
About this item
High-impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface.
- Polystyrene fabrication
- Smooth and flexible
- Sterile
Needles are straight and suitable for removal of single colony specimens. Packed sterile in safe, tamperproof, zip-seal resealable bags. Disposable inoculating loops and needles do not need flaming, eliminating the risk of infection due to aerosol formation of pathogenic substances as well as cross-contamination due to improper sterilisation. Loops and needles can be used under hoods without danger. Color-coded for easy size identification. Total length: 227 mm.